Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001320687.1 | NP_001307616.1 | 13 | 2294 | |
NM_016166.2 | NP_057250.1 | 12 | 1695 | ||
XM_011522126.2 | XP_011520428.1 | 14 | 2402 | ||
XM_011522127.2 | XP_011520429.1 | 12 | 2179 | ||
XM_017022688.1 | XP_016878177.1 | 12 | 2173 | ||
XM_017022689.1 | XP_016878178.1 | 12 | 2186 | ||
XM_017022690.1 | XP_016878179.1 | 12 | 1991 | ||
XM_017022691.1 | XP_016878180.1 | 12 | 1765 | ||
GenBank mRNA | AF077951.1 | - | 12 | 1544 | |
AF167160.1 | - | 12 | 1643 | ||
AK094641.1 | - | 13 | 2294 | ||
AK314515.1 | - | 12 | 1570 | ||
AY826819.1 | - | 12 | 1544 | ||
BC118587.1 | - | 12 | 1597 | 40085010 | |
BC121797.1 | - | 12 | 1594 | 40085007 | |
EF487997.1 | - | 12 | 1547 | ||
EF487998.1 | - | 12 | 1547 | ||
U78524.1 | - | 11 | 1529 |