Gene Transcripts
- Gene Symbol
- CRCP
- Entrez Gene ID
- 27297
- Gene Name
- CGRP receptor component
- Gene Aliases
- CGRP-RCP, CGRPRCP, RCP, RCP9
- Chromosome Location
- Chr. 7: 66114818 - 66154568 on Build GRCh38
- UniGene ID
- Hs.300684
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NR_024548.1 |
|
3, 4 |
313 |
|
GenBank mRNA |
BC052570.1 |
- |
3, 4 |
527 |
|
DA955978.1 |
- |
3, 4 |
251 |
|
View Full Product Details