Gene Transcripts
- Gene Symbol
- 4930471M09Rik
- Entrez Gene ID
- 75787
- Gene Name
- RIKEN cDNA 4930471M09 gene
- Gene Aliases
- 4930427C17Rik
- Chromosome Location
- Chr. 6: 91430460 - 91440854 on Build GRCm38
- UniGene ID
- Mm.30842
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AK015209.1 |
- |
1 |
1792 |
|
View Full Product Details