Gene Transcripts
- Gene Symbol
- Gm39168
- Entrez Gene ID
- 105243177
- Gene Name
- predicted gene, 39168
- Gene Aliases
- Chromosome Location
- Chr. 8: 46299117 - 46325729 on Build GRCm38
- UniGene ID
- Mm.114690
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AK078582.1 |
- |
5 |
1342 |
|
View Full Product Details