Gene Transcripts
- Gene Symbol
- FAM87B
- Entrez Gene ID
- 400728
- Gene Name
- family with sequence similarity 87 member B
- Gene Aliases
- Chromosome Location
- Chr. 1: 817371 - 819834 on Build GRCh38
- UniGene ID
- Hs.568078
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NR_103536.1 |
|
2 |
1593 |
|
GenBank mRNA |
AK097327.1 |
- |
2 |
1593 |
|
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
Transcript Location |
GENCODE |
ENST00000326734.2 |
- |
2 |
1593 |
|
Chr. 1 - 817371 - 819834 [+] |
View Full Product Details