Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_006509.3 | NP_006500.2 | 10 | 1350 | |
XM_005259127.3 | XP_005259184.1 | 9 | 1320 | ||
XM_005259128.2 | XP_005259185.1 | 9 | 1134 | ||
GenBank mRNA | AK290594.1 | - | 10 | 1257 | |
BC028013.1 | - | 10 | 1302 | 5215944 | |
M83221.1 | - | 10 | 1363 |