Gene Transcripts

Gene Symbol
Tdg
Entrez Gene ID
114521
Gene Name
thymine-DNA glycosylase
Gene Aliases
Chromosome Location
Chr. 7: 27194000 - 27213676 on Build Rnor_6.0
UniGene ID
Rn.98685
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_053729.1 NP_446181.1 5, 6 647
GenBank mRNA AY026945.1 - 5, 6 647
BC129088.1 - 5, 6 664 8362408

View Full Product Details