Gene Transcripts
- Gene Symbol
- Rcvrn
- Entrez Gene ID
- 140936
- Gene Name
- recoverin
- Gene Aliases
- Rcv1
- Chromosome Location
- Chr. 10: 54246250 - 54253996 on Build Rnor_6.0
- UniGene ID
- Rn.38641
- Species
- Rattus norvegicus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_080901.1 |
NP_543177.1
|
2, 3 |
494 |
|
GenBank mRNA |
AY063482.1 |
- |
2, 3 |
494 |
|
View Full Product Details