Gene Transcripts

Gene Symbol
Dusp19
Entrez Gene ID
311151
Gene Name
dual specificity phosphatase 19
Gene Aliases
Chromosome Location
Chr. 3: 67849966 - 67862475 on Build Rnor_6.0
UniGene ID
Rn.65563
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001107739.2 NP_001101209.2 1 248

View Full Product Details