Gene Transcripts
- Gene Symbol
- LOC691030
- Entrez Gene ID
- 691030
- Gene Name
- similar to RIKEN cDNA 1700001F22
- Gene Aliases
- Chromosome Location
- Chr. X: 130794079 - 130794811 on Build Rnor_6.0
- Species
- Rattus norvegicus
- Gene Symbol
- LOC100364877
- Entrez Gene ID
- 100364877
- Gene Name
- high-mobility group box 4-like
- Gene Aliases
- Chromosome Location
- Chr. X: 158694295 - 158694801 on Build Rnor_6.0
- Species
- Rattus norvegicus
- Gene Symbol
- LOC103694516
- Entrez Gene ID
- 103694516
- Gene Name
- high mobility group protein B4-like
- Gene Aliases
- Chromosome Location
- Chr. X: 134389417 - 134389923 on Build Rnor_6.0
- Species
- Rattus norvegicus
- Gene Symbol
- LOC103694587
- Entrez Gene ID
- 103694587
- Gene Name
- high mobility group protein B4-like
- Gene Aliases
- Chromosome Location
- Chr. X|NW_007906396: 20328 - 20834 on Build Rnor_6.0
- Species
- Rattus norvegicus
- Gene Symbol
- RGD1561558
- Entrez Gene ID
- 317314
- Gene Name
- similar to RIKEN cDNA 1700001F22
- Gene Aliases
- Chromosome Location
- Chr. X: 158261211 - 158261717 on Build Rnor_6.0
- Species
- Rattus norvegicus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
XM_228669.5 |
XP_228669.3
|
|
324 |
|
- Gene Symbol
- RGD1565430
- Entrez Gene ID
- 367928
- Gene Name
- similar to RIKEN cDNA 1700001F22
- Gene Aliases
- Chromosome Location
- Chr. X: 134379511 - 134383543 on Build Rnor_6.0
- Species
- Rattus norvegicus
View Full Product Details