Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_000170.2 | NP_000161.2 | 16, 17 | 2126 | |
GenBank mRNA | AK308470.1 | - | 2, 3 | 368 | |
AK310534.1 | - | 12, 13 | 1501 | ||
AK314156.1 | - | 16, 17 | 2128 | ||
BC111993.1 | - | 16, 17 | 1999 | 8327461 | |
BC111995.1 | - | 16, 17 | 1999 | 8327463 | |
D90239.1 | - | 16, 17 | 2083 | ||
M63635.1 | - | 16, 17 | 2014 | ||
M64590.1 | - | 16, 17 | 2083 |