Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_025390.4 | NP_079666.1 | 6 | 690 | |
GenBank mRNA | AK002855.1 | - | 6 | 691 | |
AK003293.2 | - | 6 | 691 | ||
AK003908.1 | - | 6 | 704 | ||
AK054466.1 | - | 723 | |||
AK150459.1 | - | 6 | 697 | ||
AK152948.1 | - | 6 | 697 |