Gene Transcripts

Gene Symbol
Tes
Entrez Gene ID
21753
Gene Name
testis derived transcript
Gene Aliases
D6Ertd352e, TESS, Tes1, Tes2, testin2
Chromosome Location
Chr. 6: 17065149 - 17105825 on Build GRCm38
UniGene ID
Mm.436548
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_207176.3 NP_997059.1 7 1868
GenBank mRNA AK132249.1 - 7 1828
AK145609.1 - 7 1817
AK167101.1 - 7 1817
AK167274.1 - 7 1826

View Full Product Details