Gene Transcripts
- Gene Symbol
- PPM1L
- Entrez Gene ID
- 151742
- Gene Name
- protein phosphatase, Mg2+/Mn2+ dependent 1L
- Gene Aliases
- PP2C-epsilon, PP2CE, PPM1-LIKE
- Chromosome Location
- Chr. 3: 160755532 - 161078907 on Build GRCh38
- UniGene ID
- Hs.389027
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AK055115.1 |
- |
5 |
2525 |
|
View Full Product Details