Gene Transcripts

Gene Symbol
NEK10
Entrez Gene ID
152110
Gene Name
NIMA related kinase 10
Gene Aliases
Chromosome Location
Chr. 3: 27107336 - 27369460 on Build GRCh38
UniGene ID
Hs.506115
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001031741.3 NP_001026911.1 12 1367
NM_001304384.1 NP_001291313.1 13 1397
NM_152534.4 NP_689747.3 35 3497
XM_006712997.3 XP_006713060.1 34 3428
XM_006712998.2 XP_006713061.1 35 3458
XM_006712999.3 XP_006713062.1 35 3458
XM_006713001.3 XP_006713064.1 33 3287
XM_017005761.1 XP_016861250.1 34 3410
XM_017005762.1 XP_016861251.1 36 3574
XM_017005763.1 XP_016861252.1 35 3555
XM_017005764.1 XP_016861253.1 34 3387
XM_017005765.1 XP_016861254.1 34 3359
XM_017005766.1 XP_016861255.1 33 3380
XM_017005767.1 XP_016861256.1 33 3323
XM_017005768.1 XP_016861257.1 34 3320
XM_017005769.1 XP_016861258.1 34 3317
XM_017005770.1 XP_016861259.1 33 3269
XM_017005771.1 XP_016861260.1 32 3239
XM_017005772.1 XP_016861261.1 32 3182
GenBank mRNA AK057247.1 - 13 1349
BC045758.1 - 12 1319 4821808
DQ104438.1 - 10 949

View Full Product Details