Gene Transcripts

Gene Symbol
FXYD3
Entrez Gene ID
5349
Gene Name
FXYD domain containing ion transport regulator 3
Gene Aliases
MAT8, PLML
Chromosome Location
Chr. 19: 35115821 - 35124324 on Build GRCh38
UniGene ID
Hs.301350
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001136007.1 NP_001129479.1 11 1393
NM_001136011.1 NP_001129483.1 10 1271
NM_001136012.1 NP_001129484.1 8 1265
NM_005971.3 NP_005962.1 9 1187
NM_021910.2 NP_068710.1 9 1349
GenBank mRNA BC090044.1 - 9 1136
U28249.1 - 9 1275
U31214.1 - 1 264

View Full Product Details