Gene Transcripts

Gene Symbol
Scn7a
Entrez Gene ID
20272
Gene Name
sodium channel, voltage-gated, type VII, alpha
Gene Aliases
1110034K09Rik, AI642000, NaG, Nav2, Nav2.3, Nax, Scn6a
Chromosome Location
Chr. 2: 66673424 - 66785091 on Build GRCm38
UniGene ID
Mm.38127
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_009135.2 NP_033161.2 2 344
GenBank mRNA AK142964.1 - 2 349
BC145273.1 - 2 201
L36179.1 - 2 380

View Full Product Details