Gene Transcripts

Gene Symbol
Ppm1h
Entrez Gene ID
319468
Gene Name
protein phosphatase 1H (PP2C domain containing)
Gene Aliases
A430075L18Rik, ARHCL1, C030002B11Rik
Chromosome Location
Chr. 10: 122678106 - 122945795 on Build GRCm38
UniGene ID
Mm.489703
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001110218.1 NP_001103688.1 3, 4 1188
NM_176919.4 NP_795893.2 3, 4 1188
GenBank mRNA AK029461.1 - 3, 4 750
AK040194.1 - 3, 4 932
AK040207.1 - 3, 4 1173
AK043121.1 - 3, 4 1189
AK051434.1 - 3, 4 1147
AK077422.1 - 3, 4 1184
AK134804.1 - 3, 4 1168
AK150309.1 - 3, 4 750
AK220260.1 - 3, 4 1145
BC052910.1 - 3, 4 499

View Full Product Details