Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_002744.4 | NP_002735.3 | 3 | 429 | |
XM_011541773.1 | XP_011540075.1 | 3 | 425 | ||
XM_011541775.2 | XP_011540077.1 | 3 | 425 | ||
XM_017001789.1 | XP_016857278.1 | 3 | 425 | ||
XM_017001790.1 | XP_016857279.1 | 3 | 425 | ||
XM_017001803.1 | XP_016857292.1 | 3 | 425 | ||
GenBank mRNA | BC008058.2 | - | 3 | 376 | |
BC014270.2 | - | 3 | 429 | 3835020 | |
BT007082.1 | - | 3 | 268 | ||
L14283.1 | - | 3 | 268 | ||
Z15108.1 | - | 3 | 250 |