Gene Transcripts

Gene Symbol
Nalcn
Entrez Gene ID
338370
Gene Name
sodium leak channel, non-selective
Gene Aliases
A530023G15Rik, AI849508, Vgcnl1
Chromosome Location
Chr. 14: 123276634 - 123627363 on Build GRCm38
UniGene ID
Mm.255348
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_177393.4 NP_796367.3 40 4903
GenBank mRNA AK044431.1 - 8 850

View Full Product Details