Gene Transcripts

Gene Symbol
Lactb2
Entrez Gene ID
212442
Gene Name
lactamase, beta 2
Gene Aliases
Cgi-83, E430032H21Rik
Chromosome Location
Chr. 1: 13625893 - 13660605 on Build GRCm38
UniGene ID
Mm.89572
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_145381.2 NP_663356.1 3 360
GenBank mRNA AK088932.1 - 3 373
AK090091.1 - 3 389
AK143912.1 - 3 339
BC004045.1 - 3 360 3496398

View Full Product Details