Gene Transcripts

Gene Symbol
Clic5
Entrez Gene ID
224796
Gene Name
chloride intracellular channel 5
Gene Aliases
5730531E12Rik, B330005L24, Gm322, jbg
Chromosome Location
Chr. 17: 44134678 - 44280168 on Build GRCm38
UniGene ID
Mm.37666
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_172621.2 NP_766209.1 6 3253
GenBank mRNA AK030659.1 - 1 1332
AK046522.1 - 6 3244
AK167584.1 - 1 1274

View Full Product Details