Gene Transcripts

Gene Symbol
Phactr1
Entrez Gene ID
218194
Gene Name
phosphatase and actin regulator 1
Gene Aliases
9630030F18Rik, Rpel1
Chromosome Location
Chr. 13: 42680571 - 43138526 on Build GRCm38
UniGene ID
Mm.160124
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001005740.1 NP_001005740.1 5, 6 889
GenBank mRNA AK048208.1 - 5, 6 785
AK139452.1 - 5, 6 861
BC061691.1 - 5, 6 889 6307669

View Full Product Details