Gene Transcripts

Gene Symbol
Srrm2
Entrez Gene ID
75956
Gene Name
serine/arginine repetitive matrix 2
Gene Aliases
5033413A03Rik, AA410130, SRm300, mKIAA0324
Chromosome Location
Chr. 17: 23790857 - 23824741 on Build GRCm38
UniGene ID
Mm.436876
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_175229.3 NP_780438.2 7, 8 712
GenBank mRNA AK028296.1 - 1, 2 54
AK090123.1 - 7, 8 673
AK171001.1 - 8, 9 985

View Full Product Details