Gene Transcripts

Gene Symbol
Wfs1
Entrez Gene ID
83725
Gene Name
Wolfram syndrome 1 (wolframin)
Gene Aliases
Chromosome Location
Chr. 14: 78640707 - 78665224 on Build Rnor_6.0
UniGene ID
Rn.229139
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_031823.1 NP_114011.1 1 237
GenBank mRNA AF136378.1 - 1 237

View Full Product Details