Gene Transcripts

Gene Symbol
Cx3cl1
Entrez Gene ID
89808
Gene Name
chemokine (C-X3-C motif) ligand 1
Gene Aliases
Cx3c, Scyd1
Chromosome Location
Chr. 19: 10644267 - 10654946 on Build Rnor_6.0
UniGene ID
Rn.107266
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_134455.1 NP_604450.1 3 568
XM_008772329.1 XP_008770551.1 4 1305
GenBank mRNA AF030358.2 - 3 568
BC070938.1 - 3 569 7099250

View Full Product Details