Gene Transcripts
- Gene Symbol
- RAB3IL1
- Entrez Gene ID
- 5866
- Gene Name
- RAB3A interacting protein like 1
- Gene Aliases
- GRAB
- Chromosome Location
- Chr. 11: 61897296 - 61946312 on Build GRCh38
- UniGene ID
- Hs.13759
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001271686.1 |
NP_001258615.1
|
3, 4 |
586 |
|
NM_013401.3 |
NP_037533.2
|
3, 4 |
610 |
|
XM_005274140.4 |
XP_005274197.1
|
3, 4 |
575 |
|
XM_006718647.3 |
XP_006718710.1
|
3, 4 |
577 |
|
XM_011545196.2 |
XP_011543498.1
|
3, 4 |
581 |
|
XM_011545197.2 |
XP_011543499.1
|
3, 4 |
897 |
|
XM_011545198.2 |
XP_011543500.1
|
3, 4 |
605 |
|
XM_011545199.2 |
XP_011543501.1
|
3, 4 |
583 |
|
XM_011545201.2 |
XP_011543503.1
|
3, 4 |
588 |
|
GenBank mRNA |
AF084557.1 |
- |
2, 3 |
164 |
|
BC022239.1 |
- |
3, 4 |
526 |
4390573 |
BC051820.1 |
- |
3, 4 |
586 |
5188805 |
BQ946081.1 |
- |
3, 4 |
552 |
|
DR004757.1 |
- |
3, 4 |
612 |
|
View Full Product Details