Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_006983.1 | NP_008914.1 | 1 | 113 | |
XM_017002615.1 | XP_016858104.1 | 1 | 613 | ||
XM_017002616.1 | XP_016858105.1 | 1 | 613 | ||
XM_017002617.1 | XP_016858106.1 | 1 | 613 | ||
GenBank mRNA | AB010961.1 | - | 1 | 102 | |
AB010962.1 | - | 1 | 127 | ||
AF056200.1 | - | 1 | 90 | ||
AJ005256.1 | - | 1 | 113 | ||
AK297700.1 | - | 1 | 117 | ||
BC025719.1 | - | 1 | 104 | 5228894 |