Gene Transcripts

Gene Symbol
NEK10
Entrez Gene ID
152110
Gene Name
NIMA related kinase 10
Gene Aliases
Chromosome Location
Chr. 3: 27107336 - 27369460 on Build GRCh38
UniGene ID
Hs.506115
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_152534.4 NP_689747.3 3 342
NM_199347.3 NP_955379.2 3 342
XM_006712997.3 XP_006713060.1 3 303
XM_006712998.2 XP_006713061.1 3 303
XM_006712999.3 XP_006713062.1 3 303
XM_006713001.3 XP_006713064.1 3 303
XM_006713002.1 XP_006713065.1 3 303
XM_011533413.2 XP_011531715.1 3 303
XM_011533414.2 XP_011531716.1 3 303
XM_011533415.1 XP_011531717.1 3 303
XM_017005761.1 XP_016861250.1 2 255
XM_017005762.1 XP_016861251.1 4 419
XM_017005763.1 XP_016861252.1 3 400
XM_017005764.1 XP_016861253.1 2 232
XM_017005765.1 XP_016861254.1 2 204
XM_017005766.1 XP_016861255.1 2 255
XM_017005767.1 XP_016861256.1 2 255
XM_017005768.1 XP_016861257.1 3 303
XM_017005769.1 XP_016861258.1 3 303
XM_017005770.1 XP_016861259.1 2 255
XM_017005771.1 XP_016861260.1 2 255
XM_017005772.1 XP_016861261.1 2 255
XM_017005773.1 XP_016861262.1 3 303
XM_017005774.1 XP_016861263.1 2 255
GenBank mRNA AK123061.1 - 3 303
AK128585.1 - 2 232
DB094082.1 - 2 303

View Full Product Details