Gene Transcripts

Gene Symbol
FUT7
Entrez Gene ID
2529
Gene Name
fucosyltransferase 7
Gene Aliases
FucT-VII
Chromosome Location
Chr. 9: 137030174 - 137032840 on Build GRCh38
UniGene ID
Hs.457
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_004479.3 NP_004470.1 2 2128
GenBank mRNA BC086312.1 - 2 2128
X78031.1 - 2 1401

View Full Product Details