Gene Transcripts

Gene Symbol
SNAP91
Entrez Gene ID
9892
Gene Name
synaptosome associated protein 91
Gene Aliases
AP180, CALM
Chromosome Location
Chr. 6: 83552885 - 83709408 on Build GRCh38
UniGene ID
Hs.368046
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001242792.1 NP_001229721.1 13 1266
NM_001242793.1 NP_001229722.1 12 1059
NM_001242794.1 NP_001229723.1 11 1017
NM_001256717.1 NP_001243646.1 15 1495
NM_001256718.1 NP_001243647.1 12 976
NM_014841.2 NP_055656.1 13 1269
XM_005248770.4 XP_005248827.1 13 1108
XM_006715615.1 XP_006715678.1 13 1060
XM_011536265.1 XP_011534567.1 13 1067
XM_011536266.1 XP_011534568.1 13 1179
XM_011536269.1 XP_011534571.1 13 1060
XM_011536271.1 XP_011534573.1 12 1018
XM_011536273.2 XP_011534575.1 13 1060
XM_011536275.2 XP_011534577.1 13 1060
XM_011536276.2 XP_011534578.1 13 1060
XM_011536277.1 XP_011534579.1 15 1348
XM_017011554.1 XP_016867043.1 12 1218
XM_017011555.1 XP_016867044.1 12 1102
XM_017011556.1 XP_016867045.1 12 1034
XM_017011557.1 XP_016867046.1 13 1237
XM_017011558.1 XP_016867047.1 13 1067
XM_017011559.1 XP_016867048.1 13 1166
XM_017011560.1 XP_016867049.1 13 1108
XM_017011561.1 XP_016867050.1 13 1179
XM_017011562.1 XP_016867051.1 13 1237
XM_017011563.1 XP_016867052.1 12 1231
XM_017011564.1 XP_016867053.1 12 1102
XM_017011565.1 XP_016867054.1 12 1160
XM_017011566.1 XP_016867055.1 12 1195
XM_017011567.1 XP_016867056.1 12 998
XM_017011568.1 XP_016867057.1 11 1189
XM_017011569.1 XP_016867058.1 12 1195
XM_017011570.1 XP_016867059.1 13 1237
XM_017011571.1 XP_016867060.1 13 1108
XM_017011572.1 XP_016867061.1 13 1067
XM_017011573.1 XP_016867062.1 13 1166
XM_017011574.1 XP_016867063.1 13 1237
XM_017011575.1 XP_016867064.1 13 1237
XM_017011576.1 XP_016867065.1 13 1040
XM_017011577.1 XP_016867066.1 13 1166
XM_017011578.1 XP_016867067.1 13 1179
XM_017011579.1 XP_016867068.1 13 1237
XM_017011580.1 XP_016867069.1 13 1237
XM_017011581.1 XP_016867070.1 13 1067
XM_017011582.1 XP_016867071.1 13 1096
XM_017011583.1 XP_016867072.1 13 1224
XM_017011584.1 XP_016867073.1 13 1067
XM_017011585.1 XP_016867074.1 13 1108
XM_017011586.1 XP_016867075.1 13 1060
XM_017011587.1 XP_016867076.1 13 1166
XM_017011588.1 XP_016867077.1 13 1179
XM_017011589.1 XP_016867078.1 12 1195
XM_017011590.1 XP_016867079.1 13 1108
GenBank mRNA AB014556.1 - 13 1195
AK289582.1 - 13 1237
AK294556.1 - 6 227
AK294898.1 - 12 960
BC060818.1 - 11 1001 4827211
CR749348.1 - 15 1462

View Full Product Details