Gene Transcripts

Gene Symbol
EARS2
Entrez Gene ID
124454
Gene Name
glutamyl-tRNA synthetase 2, mitochondrial
Gene Aliases
COXPD12, MSE1, gluRS
Chromosome Location
Chr. 16: 23522013 - 23557375 on Build GRCh38
UniGene ID
Hs.620541
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001083614.1 NP_001077083.1 9 1647
NR_003501.1 9 1647
XM_011545738.1 XP_011544040.1 9 1937
XM_011545739.1 XP_011544041.1 9 1556
GenBank mRNA AB075850.1 - 9 1604
AK095998.1 - 9 1636
AL832489.1 - 9 1623
BX648129.1 - 7 1326

View Full Product Details