Gene Transcripts

Gene Symbol
LOC100128288
Entrez Gene ID
100128288
Gene Name
uncharacterized LOC100128288
Gene Aliases
Chromosome Location
Chr. 17: 8358413 - 8360541 on Build GRCh38
UniGene ID
Hs.549913
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NR_024447.1 1 723
GenBank mRNA AK094860.1 - 1 402

View Full Product Details