Gene Transcripts

Gene Symbol
LINC01588
Entrez Gene ID
283551
Gene Name
long intergenic non-protein coding RNA 1588
Gene Aliases
C14orf182
Chromosome Location
Chr. 14: 49981669 - 50007520 on Build GRCh38
UniGene ID
Hs.292580
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NR_126496.1 1 3578
GenBank mRNA AK090420.1 - 1 3578
BC128083.1 - 2 1695 40075904

View Full Product Details