Gene Transcripts
- Gene Symbol
- LINC01588
- Entrez Gene ID
- 283551
- Gene Name
- long intergenic non-protein coding RNA 1588
- Gene Aliases
- C14orf182
- Chromosome Location
- Chr. 14: 49981669 - 50007520 on Build GRCh38
- UniGene ID
- Hs.292580
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NR_126496.1 |
|
1 |
3327 |
|
GenBank mRNA |
AK090420.1 |
- |
1 |
3327 |
|
View Full Product Details