Gene Transcripts
- Gene Symbol
- LINC00593
- Entrez Gene ID
- 414926
- Gene Name
- long intergenic non-protein coding RNA 593
- Gene Aliases
- C15orf50
- Chromosome Location
- Chr. 15: 69835234 - 69842967 on Build GRCh38
- UniGene ID
- Hs.569502
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NR_026764.1 |
|
5 |
439 |
|
GenBank mRNA |
BC031958.1 |
- |
5 |
439 |
|
View Full Product Details