Gene Transcripts

Gene Symbol
EXOC3-AS1
Entrez Gene ID
116349
Gene Name
EXOC3 antisense RNA 1
Gene Aliases
C5orf55
Chromosome Location
Chr. 5: 441835 - 443160 on Build GRCh38
UniGene ID
Hs.446702
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NR_126522.1 1 459
GenBank mRNA BC014011.2 - 1 394
BC029796.1 - 1 442 5183143

View Full Product Details