Gene Transcripts
- Gene Symbol
- PIKFYVE
- Entrez Gene ID
- 200576
- Gene Name
- phosphoinositide kinase, FYVE-type zinc finger containing
- Gene Aliases
- CFD, FAB1, HEL37, PIP5K, PIP5K3, ZFYVE29
- Chromosome Location
- Chr. 2: 208266178 - 208358751 on Build GRCh38
- UniGene ID
- Hs.744997
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001178000.1 |
NP_001171471.1
|
11, 12 |
1624 |
|
NM_015040.3 |
NP_055855.2
|
11, 12 |
1624 |
|
NM_152671.3 |
NP_689884.1
|
9, 10 |
1333 |
|
XM_011510778.2 |
XP_011509080.1
|
12, 13 |
1674 |
|
XM_011510779.2 |
XP_011509081.1
|
12, 13 |
1607 |
|
XM_011510780.2 |
XP_011509082.1
|
12, 13 |
1692 |
|
XM_011510782.2 |
XP_011509084.1
|
12, 13 |
1674 |
|
XM_011510786.2 |
XP_011509088.1
|
10, 11 |
1341 |
|
XM_011510787.1 |
XP_011509089.1
|
10, 11 |
1350 |
|
XM_011510788.1 |
XP_011509090.1
|
9, 10 |
1333 |
|
XM_011510789.2 |
XP_011509091.1
|
8, 9 |
1155 |
|
XM_011510790.1 |
XP_011509092.1
|
10, 11 |
1452 |
|
XM_011510792.2 |
XP_011509094.1
|
12, 13 |
1674 |
|
XM_017003572.1 |
XP_016859061.1
|
10, 11 |
1328 |
|
XM_017003573.1 |
XP_016859062.1
|
7, 8 |
1036 |
|
XM_017003574.1 |
XP_016859063.1
|
6, 7 |
707 |
|
GenBank mRNA |
AY457063.1 |
- |
11, 12 |
1628 |
|
BC017736.2 |
- |
10, 11 |
1452 |
|
BC032389.1 |
- |
9, 10 |
1333 |
|
BC125052.1 |
- |
10, 11 |
1355 |
|
BC125053.1 |
- |
11, 12 |
1613 |
|
EU794609.1 |
- |
11, 12 |
1628 |
|
View Full Product Details