Gene Transcripts
- Gene Symbol
- NOXO1
- Entrez Gene ID
- 124056
- Gene Name
- NADPH oxidase organizer 1
- Gene Aliases
- P41NOX, P41NOXA, P41NOXB, P41NOXC, SH3PXD5, SNX28
- Chromosome Location
- Chr. 16: 1978917 - 1982751 on Build GRCh38
- UniGene ID
- Hs.191762
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001267721.1 |
NP_001254650.1
|
1, 2 |
429 |
|
NM_144603.3 |
NP_653204.1
|
1, 2 |
429 |
|
NM_172167.2 |
NP_751907.1
|
1, 2 |
429 |
|
NM_172168.2 |
NP_751908.1
|
1, 2 |
429 |
|
XM_005255099.4 |
XP_005255156.1
|
2, 3 |
1524 |
|
XM_017022927.1 |
XP_016878416.1
|
2, 3 |
1030 |
|
XM_017022928.1 |
XP_016878417.1
|
2, 3 |
387 |
|
GenBank mRNA |
AB097667.1 |
- |
1, 2 |
239 |
|
AF532983.1 |
- |
1, 2 |
63 |
|
AF532984.1 |
- |
1, 2 |
63 |
|
AF532985.1 |
- |
1, 2 |
63 |
|
AF539796.1 |
- |
1, 2 |
59 |
|
AK292197.1 |
- |
1, 2 |
429 |
|
AK310198.1 |
- |
1, 2 |
313 |
|
AK313917.1 |
- |
1, 2 |
349 |
|
AY191359.1 |
- |
1, 2 |
63 |
|
AY255768.1 |
- |
1, 2 |
59 |
|
BC015917.2 |
- |
1, 2 |
319 |
4661469 |
DB009385.1 |
- |
1, 2 |
429 |
|
View Full Product Details