Gene Transcripts
- Gene Symbol
- IZUMO4
- Entrez Gene ID
- 113177
- Gene Name
- IZUMO family member 4
- Gene Aliases
- C19orf36, IMAGE:4215339
- Chromosome Location
- Chr. 19: 2096848 - 2099590 on Build GRCh38
- UniGene ID
- Hs.424049
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001031735.2 |
NP_001026905.2
|
1, 2 |
297 |
|
NM_001039846.1 |
NP_001034935.1
|
1, 2 |
297 |
|
XM_005259480.3 |
XP_005259537.2
|
1, 2 |
318 |
|
GenBank mRNA |
AK294083.1 |
- |
1, 2 |
168 |
|
AK301957.1 |
- |
1, 2 |
251 |
|
AY358444.1 |
- |
1, 2 |
297 |
|
AY753639.1 |
- |
1, 2 |
256 |
|
BC010443.1 |
- |
1, 2 |
236 |
|
BC014609.2 |
- |
1, 2 |
251 |
4825119 |
BC040964.1 |
- |
1, 2 |
238 |
|
BI827974.1 |
- |
1, 2 |
154 |
|
BX378854.2 |
- |
1, 2 |
297 |
|
DQ196309.1 |
- |
1, 2 |
284 |
|
DQ196310.1 |
- |
1, 2 |
284 |
|
DQ424900.1 |
- |
1, 2 |
220 |
|
View Full Product Details