Gene Transcripts
- Gene Symbol
- ASMTL-AS1
- Entrez Gene ID
- 80161
- Gene Name
- ASMTL antisense RNA 1
- Gene Aliases
- ASMTL-AS, ASMTLAS, CXYorf2, NCRNA00105
- Chromosome Location
- Chr. Y: 1400531 - 1415421 on Build GRCh38
- UniGene ID
- Hs.521856
- Species
- Homo sapiens
- Gene Symbol
- ASMTL
- Entrez Gene ID
- 8623
- Gene Name
- acetylserotonin O-methyltransferase-like
- Gene Aliases
- ASMTLX, ASMTLY, ASTML
- Chromosome Location
- Chr. X: 1403139 - 1453794 on Build GRCh38
- UniGene ID
- Hs.533514
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
BU688748.1 |
- |
2 |
436 |
|
View Full Product Details