Gene Transcripts

Gene Symbol
PPP6R3
Entrez Gene ID
55291
Gene Name
protein phosphatase 6 regulatory subunit 3
Gene Aliases
C11orf23, PP6R3, SAP190, SAPL, SAPLa, SAPS3
Chromosome Location
Chr. 11: 68460712 - 68615334 on Build GRCh38
UniGene ID
Hs.503022
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001164160.1 NP_001157632.1 13, 14 1721
NM_001164161.1 NP_001157633.1 13, 14 1721
NM_001164162.1 NP_001157634.1 13, 14 1721
NM_001164163.1 NP_001157635.1 13, 14 1721
NM_001164164.1 NP_001157636.1 13, 14 1673
NM_018312.4 NP_060782.2 13, 14 1673
XM_006718608.3 XP_006718671.1 14, 15 1837
XM_006718612.3 XP_006718675.1 14, 15 1837
XM_006718613.2 XP_006718676.1 14, 15 1837
XM_006718614.2 XP_006718677.1 14, 15 1837
XM_006718618.3 XP_006718681.1 14, 15 1837
XM_006718621.2 XP_006718684.1 14, 15 1837
XM_006718622.3 XP_006718685.1 13, 14 1684
XM_006718623.3 XP_006718686.1 13, 14 1667
XM_006718624.2 XP_006718687.1 13, 14 1684
XM_006718627.3 XP_006718690.1 14, 15 1837
XM_011545135.1 XP_011543437.1 14, 15 1837
XM_011545136.2 XP_011543438.1 13, 14 1720
XM_011545137.2 XP_011543439.1 14, 15 1820
XM_011545138.2 XP_011543440.1 15, 16 2038
XM_011545139.1 XP_011543441.1 14, 15 1837
XM_011545140.2 XP_011543442.1 14, 15 1837
XM_011545141.1 XP_011543443.1 14, 15 1837
XM_011545142.2 XP_011543444.1 14, 15 1837
XM_011545143.1 XP_011543445.1 14, 15 1837
XM_011545145.2 XP_011543447.1 13, 14 1684
XM_017017962.1 XP_016873451.1 13, 14 1675
XM_017017963.1 XP_016873452.1 14, 15 1761
XM_017017964.1 XP_016873453.1 14, 15 1837
XM_017017965.1 XP_016873454.1 13, 14 1720
XM_017017966.1 XP_016873455.1 14, 15 1923
XM_017017967.1 XP_016873456.1 14, 15 1837
XM_017017968.1 XP_016873457.1 13, 14 1720
XM_017017969.1 XP_016873458.1 14, 15 1837
XM_017017970.1 XP_016873459.1 13, 14 1720
XM_017017971.1 XP_016873460.1 14, 15 1837
XM_017017972.1 XP_016873461.1 13, 14 1693
XM_017017973.1 XP_016873462.1 14, 15 1820
XM_017017974.1 XP_016873463.1 14, 15 1837
XM_017017975.1 XP_016873464.1 14, 15 1837
XM_017017976.1 XP_016873465.1 12, 13 1567
XM_017017977.1 XP_016873466.1 13, 14 1608
XM_017017978.1 XP_016873467.1 12, 13 1459
XM_017017979.1 XP_016873468.1 12, 13 1459
XM_017017980.1 XP_016873469.1 12, 13 1459
XM_017017981.1 XP_016873470.1 12, 13 1459
XM_017017982.1 XP_016873471.1 12, 13 1459
XM_017017983.1 XP_016873472.1 13, 14 1858
XM_017017984.1 XP_016873473.1 13, 14 1788
GenBank mRNA AB046778.2 - 13, 14 1669
AF264779.1 - 12, 13 1578
AF264780.1 - 12, 13 1578
AK057250.1 - 7, 8 794
AK300359.1 - 6, 7 642
AK302611.1 - 6, 7 662
AK303315.1 - 8, 9 1037
AK315136.1 - 13, 14 1690
BC105933.1 - 12, 13 1589 40029064
BC105934.1 - 12, 13 1589 40029058
BC107599.1 - 12, 13 1589 40029065
CR749815.1 - 13, 14 1684
CR933658.1 - 13, 14 1684
CR936718.1 - 13, 14 1689
DQ111954.2 - 11, 12 1454

View Full Product Details