Gene Transcripts
- Gene Symbol
- AP1S2
- Entrez Gene ID
- 8905
- Gene Name
- adaptor related protein complex 1 sigma 2 subunit
- Gene Aliases
- DC22, MRX59, MRXS21, MRXS5, MRXSF, PGS, SIGMA1B
- Chromosome Location
- Chr. X: 15825806 - 15855014 on Build GRCh38
- UniGene ID
- Hs.121592
- Species
- Homo sapiens
- Gene Symbol
- LOC653653
- Entrez Gene ID
- 653653
- Gene Name
- adaptor related protein complex 1 sigma 2 subunit pseudogene
- Gene Aliases
- Chromosome Location
- Chr. 17: 60101760 - 60102919 on Build GRCh38
- UniGene ID
- Hs.631741
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NR_027408.1 |
|
1 |
422 |
|
GenBank mRNA |
AK295685.1 |
- |
1 |
422 |
|
View Full Product Details