Gene Transcripts

Gene Symbol
TSPY3
Entrez Gene ID
728137
Gene Name
testis specific protein, Y-linked 3
Gene Aliases
CT78
Chromosome Location
Chr. Y: 9398421 - 9401224 on Build GRCh38
UniGene ID
Hs.556121
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001077697.2 NP_001071165.2 5 912
XM_005272742.4 XP_005272799.1 5 898
XM_011544004.2 XP_011542306.1 5 880

Gene Symbol
TSPY1
Entrez Gene ID
7258
Gene Name
testis specific protein, Y-linked 1
Gene Aliases
CT78, DYS14, TSPY, pJA923
Chromosome Location
Chr. Y: 9466955 - 9469756 on Build GRCh38
UniGene ID
Hs.556121
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001197242.2 NP_001184171.1 5 923
NM_001320964.1 NP_001307893.1 6 651
NM_003308.4 NP_003299.2 5 912
XM_017030064.1 XP_016885553.1 5 880
GenBank mRNA BC121113.1 - 6 655
M98525.1 - 5 827
U58096.1 - 5 911

Gene Symbol
TSPY10
Entrez Gene ID
100289087
Gene Name
testis specific protein, Y-linked 10
Gene Aliases
Chromosome Location
Chr. Y: 9527880 - 9530676 on Build GRCh38
UniGene ID
Hs.556121
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001282469.2 NP_001269398.1 5 912
NM_001320962.1 NP_001307891.1 5 923
XM_017030025.1 XP_016885514.1 5 880
GenBank mRNA AK308271.1 - 3 867
AY130858.1 - 5 877
BC075016.2 - 5 893 30915406
BC121114.1 - 5 916 40120435
DB545400.1 - 3 246

Gene Symbol
TSPY2
Entrez Gene ID
64591
Gene Name
testis specific protein, Y-linked 2
Gene Aliases
TSPYQ1
Chromosome Location
Chr. Y: 6246223 - 6249020 on Build GRCh38
UniGene ID
Hs.646249
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_022573.2 NP_072095.2 5 912
XM_005262569.4 XP_005262626.1 5 883
XM_017030062.1 XP_016885551.1 5 865
GenBank mRNA AK307425.1 - 5 817

Gene Symbol
TSPY4
Entrez Gene ID
728395
Gene Name
testis specific protein, Y-linked 4
Gene Aliases
TSPY10, TSPY8
Chromosome Location
Chr. Y: 9337489 - 9340285 on Build GRCh38
UniGene ID
Hs.556121
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001164471.1 NP_001157943.1 5 884
XM_005272743.4 XP_005272800.1 5 916
XM_017030065.1 XP_016885554.1 5 898
GenBank mRNA AK093413.1 - 3 1091
BX281192.1 - 4 378

Gene Symbol
TSPY8
Entrez Gene ID
728403
Gene Name
testis specific protein, Y-linked 8
Gene Aliases
Chromosome Location
Chr. Y: 9357829 - 9360600 on Build GRCh38
UniGene ID
Hs.740264
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001243721.1 NP_001230650.1 5 866
XM_005272744.4 XP_005272801.1 5 891
XM_011544005.2 XP_011542307.1 5 873

View Full Product Details