Gene Transcripts

Gene Symbol
LOC653653
Entrez Gene ID
653653
Gene Name
adaptor related protein complex 1 sigma 2 subunit pseudogene
Gene Aliases
Chromosome Location
Chr. 17: 60101760 - 60102919 on Build GRCh38
UniGene ID
Hs.631741
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NR_027408.1 1 322
GenBank mRNA AK295685.1 - 1 322

View Full Product Details