Gene Transcripts
- Gene Symbol
- SVOP
- Entrez Gene ID
- 55530
- Gene Name
- SV2 related protein
- Gene Aliases
- Chromosome Location
- Chr. 12: 108908129 - 109021240 on Build GRCh38
- UniGene ID
- Hs.27226
- Species
- Homo sapiens
- Gene Symbol
- ACTR3C
- Entrez Gene ID
- 653857
- Gene Name
- ARP3 actin-related protein 3 homolog C
- Gene Aliases
- ARP11
- Chromosome Location
- Chr. 7: 149881359 - 150325523 on Build GRCh38
- UniGene ID
- Hs.664579
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
BX640643.1 |
- |
6 |
2823 |
|
- Gene Symbol
- AGMAT
- Entrez Gene ID
- 79814
- Gene Name
- agmatinase
- Gene Aliases
- Chromosome Location
- Chr. 1: 15571699 - 15585110 on Build GRCh38
- UniGene ID
- Hs.461532
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_024758.4 |
NP_079034.3
|
7 |
2634 |
|
- Gene Symbol
- AMD1
- Entrez Gene ID
- 262
- Gene Name
- adenosylmethionine decarboxylase 1
- Gene Aliases
- ADOMETDC, AMD, SAMDC
- Chromosome Location
- Chr. 6: 110814621 - 110895713 on Build GRCh38
- UniGene ID
- Hs.159118
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AA808049.1 |
- |
1 |
262 |
|
- Gene Symbol
- ATP13A4
- Entrez Gene ID
- 84239
- Gene Name
- ATPase 13A4
- Gene Aliases
- Chromosome Location
- Chr. 3: 193398967 - 193581725 on Build GRCh38
- UniGene ID
- Hs.674423
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_032279.3 |
NP_115655.2
|
30 |
6144 |
|
- Gene Symbol
- BCO2
- Entrez Gene ID
- 83875
- Gene Name
- beta-carotene oxygenase 2
- Gene Aliases
- B-DIOX-II, BCDO2
- Chromosome Location
- Chr. 11: 112175485 - 112218929 on Build GRCh38
- UniGene ID
- Hs.728325
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AK091783.1 |
- |
8 |
2328 |
|
- Gene Symbol
- BDH1
- Entrez Gene ID
- 622
- Gene Name
- 3-hydroxybutyrate dehydrogenase, type 1
- Gene Aliases
- BDH, SDR9C1
- Chromosome Location
- Chr. 3: 197509783 - 197573323 on Build GRCh38
- UniGene ID
- Hs.274539
- Species
- Homo sapiens
- Gene Symbol
- BRCA1
- Entrez Gene ID
- 672
- Gene Name
- BRCA1, DNA repair associated
- Gene Aliases
- BRCAI, BRCC1, BROVCA1, FANCS, IRIS, PNCA4, PPP1R53, PSCP, RNF53
- Chromosome Location
- Chr. 17: 43044295 - 43125483 on Build GRCh38
- UniGene ID
- Hs.194143
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AB621825.1 |
- |
1 |
608 |
|
- Gene Symbol
- CD27-AS1
- Entrez Gene ID
- 678655
- Gene Name
- CD27 antisense RNA 1
- Gene Aliases
- Chromosome Location
- Chr. 12: 6439001 - 6451517 on Build GRCh38
- UniGene ID
- Hs.655224
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
BC041960.1 |
- |
1 |
145 |
|
BC047072.1 |
- |
3 |
687 |
|
- Gene Symbol
- CHD7
- Entrez Gene ID
- 55636
- Gene Name
- chromodomain helicase DNA binding protein 7
- Gene Aliases
- CRG, HH5, IS3, KAL5
- Chromosome Location
- Chr. 8: 60678744 - 60868028 on Build GRCh38
- UniGene ID
- Hs.20395
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AL833190.1 |
- |
1 |
2121 |
|
- Gene Symbol
- CREB3L1
- Entrez Gene ID
- 90993
- Gene Name
- cAMP responsive element binding protein 3 like 1
- Gene Aliases
- OASIS
- Chromosome Location
- Chr. 11: 46277638 - 46321422 on Build GRCh38
- UniGene ID
- Hs.405961
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AF055009.1 |
- |
1 |
171 |
|
- Gene Symbol
- EPHA7
- Entrez Gene ID
- 2045
- Gene Name
- EPH receptor A7
- Gene Aliases
- EHK-3, EHK3, EK11, HEK11
- Chromosome Location
- Chr. 6: 93240020 - 93419687 on Build GRCh38
- UniGene ID
- Hs.73962
- Species
- Homo sapiens
- Gene Symbol
- GPR137B
- Entrez Gene ID
- 7107
- Gene Name
- G protein-coupled receptor 137B
- Gene Aliases
- TM7SF1
- Chromosome Location
- Chr. 1: 236142502 - 236211059 on Build GRCh38
- UniGene ID
- Hs.498160
- Species
- Homo sapiens
- Gene Symbol
- IL13RA1
- Entrez Gene ID
- 3597
- Gene Name
- interleukin 13 receptor subunit alpha 1
- Gene Aliases
- CD213A1, CT19, IL-13Ra, NR4
- Chromosome Location
- Chr. X: 118726954 - 118794533 on Build GRCh38
- UniGene ID
- Hs.496646
- Species
- Homo sapiens
- Gene Symbol
- KIT
- Entrez Gene ID
- 3815
- Gene Name
- KIT proto-oncogene receptor tyrosine kinase
- Gene Aliases
- C-Kit, CD117, PBT, SCFR
- Chromosome Location
- Chr. 4: 54657928 - 54740715 on Build GRCh38
- UniGene ID
- Hs.479754
- Species
- Homo sapiens
- Gene Symbol
- KLHL22
- Entrez Gene ID
- 84861
- Gene Name
- kelch like family member 22
- Gene Aliases
- KELCHL
- Chromosome Location
- Chr. 22: 20441519 - 20496249 on Build GRCh38
- UniGene ID
- Hs.517419
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AK024369.1 |
- |
1 |
474 |
|
- Gene Symbol
- KLHL3
- Entrez Gene ID
- 26249
- Gene Name
- kelch like family member 3
- Gene Aliases
- PHA2D
- Chromosome Location
- Chr. 5: 137617500 - 137736090 on Build GRCh38
- UniGene ID
- Hs.655084
- Species
- Homo sapiens
- Gene Symbol
- LEMD2
- Entrez Gene ID
- 221496
- Gene Name
- LEM domain containing 2
- Gene Aliases
- CTRCT42, NET25, dJ482C21.1
- Chromosome Location
- Chr. 6: 33771213 - 33794274 on Build GRCh38
- UniGene ID
- Hs.444845
- Species
- Homo sapiens
- Gene Symbol
- LIN28A
- Entrez Gene ID
- 79727
- Gene Name
- lin-28 homolog A
- Gene Aliases
- CSDD1, LIN-28, LIN28, ZCCHC1, lin-28A
- Chromosome Location
- Chr. 1: 26410768 - 26429728 on Build GRCh38
- UniGene ID
- Hs.86154
- Species
- Homo sapiens
- Gene Symbol
- LINC00488
- Entrez Gene ID
- 677779
- Gene Name
- long intergenic non-protein coding RNA 488
- Gene Aliases
- C3orf66
- Chromosome Location
- Chr. 3: 109178165 - 109185261 on Build GRCh38
- UniGene ID
- Hs.618669
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NR_026767.1 |
|
3 |
1690 |
|
GenBank mRNA |
AK123226.1 |
- |
3 |
1690 |
|
BC022563.1 |
- |
3 |
1679 |
|
- Gene Symbol
- LINC01296
- Entrez Gene ID
- 642477
- Gene Name
- long intergenic non-protein coding RNA 1296
- Gene Aliases
- Chromosome Location
- Chr. 14: 19062377 - 19107497 on Build GRCh38
- UniGene ID
- Hs.474095
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NR_122111.1 |
|
4 |
2510 |
|
GenBank mRNA |
BC041856.1 |
- |
4 |
2487 |
|
- Gene Symbol
- LINC01299
- Entrez Gene ID
- 286186
- Gene Name
- long intergenic non-protein coding RNA 1299
- Gene Aliases
- Chromosome Location
- Chr. 8: 65527008 - 65562666 on Build GRCh38
- UniGene ID
- Hs.449427
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NR_033893.1 |
|
5 |
2500 |
|
GenBank mRNA |
AK094960.1 |
- |
5 |
2486 |
|
- Gene Symbol
- LOC100131257
- Entrez Gene ID
- 100131257
- Gene Name
- zinc finger protein 655 pseudogene
- Gene Aliases
- Chromosome Location
- Chr. 7: 7075770 - 7096786 on Build GRCh38
- UniGene ID
- Hs.551110
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NR_034022.1 |
|
1 |
20342 |
|
- Gene Symbol
- LOC100289455
- Entrez Gene ID
- 100289455
- Gene Name
- uncharacterized LOC100289455
- Gene Aliases
- Chromosome Location
- Chr. 10: 22437086 - 22438509 on Build GRCh38
- UniGene ID
- Hs.729523
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AK295718.1 |
- |
1 |
1132 |
|
- Gene Symbol
- LOC105373133
- Entrez Gene ID
- 105373133
- Gene Name
- uncharacterized LOC105373133
- Gene Aliases
- Chromosome Location
- Chr. X: 13044544 - 13086975 on Build GRCh38
- Species
- Homo sapiens
- Gene Symbol
- LOC105374290
- Entrez Gene ID
- 105374290
- Gene Name
- uncharacterized LOC105374290
- Gene Aliases
- Chromosome Location
- Chr. 3: 194499007 - 194527436 on Build GRCh38
- UniGene ID
- Hs.606585
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AK091192.1 |
- |
2 |
785 |
|
- Gene Symbol
- LOC728158
- Entrez Gene ID
- 728158
- Gene Name
- hCG2044975
- Gene Aliases
- Chromosome Location
- Chr. 10: 126423107 - 126440535 on Build GRCh38
- UniGene ID
- Hs.646701
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AK125527.1 |
- |
2 |
247 |
|
- Gene Symbol
- LRRC74B
- Entrez Gene ID
- 400891
- Gene Name
- leucine rich repeat containing 74B
- Gene Aliases
- Chromosome Location
- Chr. 22: 21045960 - 21064168 on Build GRCh38
- UniGene ID
- Hs.291198
- Species
- Homo sapiens
- Gene Symbol
- MAGI1
- Entrez Gene ID
- 9223
- Gene Name
- membrane associated guanylate kinase, WW and PDZ domain containing 1
- Gene Aliases
- AIP-3, AIP3, BAIAP1, BAP-1, BAP1, MAGI-1, Magi1d, TNRC19, WWP3
- Chromosome Location
- Chr. 3: 65353525 - 66038836 on Build GRCh38
- UniGene ID
- Hs.476636
- Species
- Homo sapiens
- Gene Symbol
- MAP9
- Entrez Gene ID
- 79884
- Gene Name
- microtubule associated protein 9
- Gene Aliases
- ASAP
- Chromosome Location
- Chr. 4: 155342658 - 155377118 on Build GRCh38
- UniGene ID
- Hs.61271
- Species
- Homo sapiens
- Gene Symbol
- MICAL3
- Entrez Gene ID
- 57553
- Gene Name
- microtubule associated monooxygenase, calponin and LIM domain containing 3
- Gene Aliases
- MICAL-3
- Chromosome Location
- Chr. 22: 17787649 - 18024559 on Build GRCh38
- UniGene ID
- Hs.528024
- Species
- Homo sapiens
- Gene Symbol
- NECTIN3
- Entrez Gene ID
- 25945
- Gene Name
- nectin cell adhesion molecule 3
- Gene Aliases
- CD113, CDW113, NECTIN-3, PPR3, PRR3, PVRL3, PVRR3
- Chromosome Location
- Chr. 3: 111071759 - 111201444 on Build GRCh38
- UniGene ID
- Hs.293917
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AA555086.1 |
- |
1 |
122 |
|
- Gene Symbol
- NKTR
- Entrez Gene ID
- 4820
- Gene Name
- natural killer cell triggering receptor
- Gene Aliases
- p104
- Chromosome Location
- Chr. 3: 42600655 - 42648741 on Build GRCh38
- UniGene ID
- Hs.529509
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
BX538094.1 |
- |
|
6798 |
|
- Gene Symbol
- NUAK1
- Entrez Gene ID
- 9891
- Gene Name
- NUAK family kinase 1
- Gene Aliases
- ARK5
- Chromosome Location
- Chr. 12: 106063347 - 106140033 on Build GRCh38
- UniGene ID
- Hs.524692
- Species
- Homo sapiens
- Gene Symbol
- OPA3
- Entrez Gene ID
- 80207
- Gene Name
- optic atrophy 3 (autosomal recessive, with chorea and spastic paraplegia)
- Gene Aliases
- MGA3
- Chromosome Location
- Chr. 19: 45527767 - 45584864 on Build GRCh38
- UniGene ID
- Hs.466945
- Species
- Homo sapiens
- Gene Symbol
- PARVA
- Entrez Gene ID
- 55742
- Gene Name
- parvin alpha
- Gene Aliases
- CH-ILKBP, MXRA2
- Chromosome Location
- Chr. 11: 12376441 - 12535356 on Build GRCh38
- UniGene ID
- Hs.432914
- Species
- Homo sapiens
- Gene Symbol
- PMS2
- Entrez Gene ID
- 5395
- Gene Name
- PMS1 homolog 2, mismatch repair system component
- Gene Aliases
- HNPCC4, MLH4, PMS2CL, PMSL2
- Chromosome Location
- Chr. 7: 5970925 - 6009106 on Build GRCh38
- UniGene ID
- Hs.632637
- Species
- Homo sapiens
- Gene Symbol
- PRDM14
- Entrez Gene ID
- 63978
- Gene Name
- PR domain 14
- Gene Aliases
- PFM11
- Chromosome Location
- Chr. 8: 70051126 - 70071327 on Build GRCh38
- UniGene ID
- Hs.736037
- Species
- Homo sapiens
- Gene Symbol
- PRKXP1
- Entrez Gene ID
- 441733
- Gene Name
- protein kinase, X-linked, pseudogene 1
- Gene Aliases
- Chromosome Location
- Chr. 15: 100547752 - 100559283 on Build GRCh38
- UniGene ID
- Hs.12250
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AK095793.1 |
- |
1 |
967 |
|
- Gene Symbol
- PRR26
- Entrez Gene ID
- 414235
- Gene Name
- proline rich 26
- Gene Aliases
- C10orf108
- Chromosome Location
- Chr. 10: 649948 - 665169 on Build GRCh38
- UniGene ID
- Hs.587103
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NR_027152.2 |
|
3 |
2770 |
|
GenBank mRNA |
BC042561.1 |
- |
3 |
2580 |
|
- Gene Symbol
- RHBDD3
- Entrez Gene ID
- 25807
- Gene Name
- rhomboid domain containing 3
- Gene Aliases
- C22orf3, HS984G1A, PTAG
- Chromosome Location
- Chr. 22: 29259855 - 29268196 on Build GRCh38
- UniGene ID
- Hs.106730
- Species
- Homo sapiens
- Gene Symbol
- ROR1-AS1
- Entrez Gene ID
- 101927034
- Gene Name
- ROR1 antisense RNA 1
- Gene Aliases
- Chromosome Location
- Chr. 1: 64094442 - 64113791 on Build GRCh38
- UniGene ID
- Hs.680824
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NR_110666.1 |
|
6 |
1437 |
|
GenBank mRNA |
BC040909.1 |
- |
6 |
1437 |
|
- Gene Symbol
- S1PR3
- Entrez Gene ID
- 1903
- Gene Name
- sphingosine-1-phosphate receptor 3
- Gene Aliases
- EDG-3, EDG3, LPB3, S1P3
- Chromosome Location
- Chr. 9: 88991409 - 89005155 on Build GRCh38
- UniGene ID
- Hs.585118
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AL832194.1 |
- |
1 |
1079 |
|
- Gene Symbol
- SIRT3
- Entrez Gene ID
- 23410
- Gene Name
- sirtuin 3
- Gene Aliases
- SIR2L3
- Chromosome Location
- Chr. 11: 215030 - 236950 on Build GRCh38
- UniGene ID
- Hs.716456
- Species
- Homo sapiens
- Gene Symbol
- SMC2-AS1
- Entrez Gene ID
- 101928550
- Gene Name
- SMC2 antisense RNA 1 (head to head)
- Gene Aliases
- Chromosome Location
- Chr. 9: 104080024 - 104092474 on Build GRCh38
- UniGene ID
- Hs.144722
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NR_121580.1 |
|
3 |
4621 |
|
GenBank mRNA |
AK125887.1 |
- |
3 |
4617 |
|
- Gene Symbol
- SPDYE16
- Entrez Gene ID
- 102723555
- Gene Name
- speedy/RINGO cell cycle regulator family member E16
- Gene Aliases
- Chromosome Location
- Chr. 7: 76532626 - 76540660 on Build GRCh38
- Species
- Homo sapiens
- Gene Symbol
- SWAP70
- Entrez Gene ID
- 23075
- Gene Name
- SWAP switching B-cell complex 70kDa subunit
- Gene Aliases
- HSPC321, SWAP-70
- Chromosome Location
- Chr. 11: 9664077 - 9752991 on Build GRCh38
- UniGene ID
- Hs.153026
- Species
- Homo sapiens
- Gene Symbol
- TENM1
- Entrez Gene ID
- 10178
- Gene Name
- teneurin transmembrane protein 1
- Gene Aliases
- ODZ1, ODZ3, TEN-M1, TNM, TNM1
- Chromosome Location
- Chr. X: 124374680 - 125204305 on Build GRCh38
- UniGene ID
- Hs.23796
- Species
- Homo sapiens
- Gene Symbol
- TRMO
- Entrez Gene ID
- 51531
- Gene Name
- tRNA methyltransferase O
- Gene Aliases
- C9orf156, HSPC219, NAP1
- Chromosome Location
- Chr. 9: 97876321 - 97922570 on Build GRCh38
- UniGene ID
- Hs.9196
- Species
- Homo sapiens
- Gene Symbol
- WDR92
- Entrez Gene ID
- 116143
- Gene Name
- WD repeat domain 92
- Gene Aliases
- Chromosome Location
- Chr. 2: 68130149 - 68157560 on Build GRCh38
- UniGene ID
- Hs.631877
- Species
- Homo sapiens
- Gene Symbol
- XIRP2-AS1
- Entrez Gene ID
- 100874011
- Gene Name
- XIRP2 antisense RNA 1
- Gene Aliases
- Chromosome Location
- Chr. 2: 167122809 - 167140955 on Build GRCh38
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NR_046665.1 |
|
2 |
1167 |
|
- Gene Symbol
- ZNF169
- Entrez Gene ID
- 169841
- Gene Name
- zinc finger protein 169
- Gene Aliases
- Chromosome Location
- Chr. 9: 94259266 - 94303967 on Build GRCh38
- UniGene ID
- Hs.387623
- Species
- Homo sapiens
View Full Product Details