Gene Transcripts

Gene Symbol
MGC116121
Entrez Gene ID
498830
Gene Name
similar to RIKEN cDNA 2700062C07
Gene Aliases
Chromosome Location
Chr. 18: 16438012 - 16447011 on Build Rnor_6.0
UniGene ID
Rn.204568
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001024905.1 NP_001020076.1 3 264
XM_008771953.1 XP_008770175.1 3 343
GenBank mRNA BC097984.1 - 3 264

View Full Product Details