Gene Transcripts

Gene Symbol
PIKFYVE
Entrez Gene ID
200576
Gene Name
phosphoinositide kinase, FYVE-type zinc finger containing
Gene Aliases
CFD, FAB1, HEL37, PIP5K, PIP5K3, ZFYVE29
Chromosome Location
Chr. 2: 208266178 - 208358751 on Build GRCh38
UniGene ID
Hs.744997
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001178000.1 NP_001171471.1 12 2091
NM_152671.3 NP_689884.1 10 1800
GenBank mRNA BC032389.1 - 10 1800

View Full Product Details