Gene Transcripts

Gene Symbol
Tat
Entrez Gene ID
24813
Gene Name
tyrosine aminotransferase
Gene Aliases
Chromosome Location
Chr. 19: 41675639 - 41686195 on Build Rnor_6.0
UniGene ID
Rn.9947
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_012668.2 NP_036800.1 4 412
GenBank mRNA BC089813.1 - 4 412
M18340.1 - 4 465
X02741.1 - 4 465

View Full Product Details