Gene Transcripts

Gene Symbol
Ccdc167
Entrez Gene ID
68597
Gene Name
coiled-coil domain containing 167
Gene Aliases
1110021J02Rik
Chromosome Location
Chr. 17: 29695977 - 29717017 on Build GRCm38
UniGene ID
Mm.390979
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001163741.2 NP_001157213.1 5 3940

View Full Product Details